4-Cyanostyrene 97% stabilized

Supplier: Thermo Scientific Chemicals
Warning


Stabilization: Stabilised with 4-tert-butylpyrocatechol (TBC) 0.05%

B24395-03 B24395-06 B24395-14
CAAAB24395-03EA 158 CAD
CAAAB24395-03 CAAAB24395-06 CAAAB24395-14
4-Cyanostyrene 97% stabilized
4-Cyanostyrene

Stab. with 0.05% 4-tert-butyl catechol


Purity: 97% CAS No.: 3435-51-6 Molecular Formula: C9H7N Formula Weight: 129.16 Storage Temperature: 2°C to 8°C Refractive Index: 1.5775 Boiling Point: 92-93°/3mm UN No.: 3276 MDL No.: MFCD00080445

Formula: C₉H₇N
Boiling Pt: 92…93 °C/3 mm
Storage Temperature: Ambient
MDL Number: MFCD00080445
CAS Number: 3435-51-6
UN: 3276
ADR: "6.1",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR