Methyl-3-bromo-4-methylbenzoate 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Methyl 3-bromo-4-methylbenzoate
B24372-06
B24372-14
B24372-22
CAAAB24372-06EA
68.1
CAD
CAAAB24372-06
CAAAB24372-14
CAAAB24372-22
Methyl-3-bromo-4-methylbenzoate 99%
Methyl 3-bromo-4-methylbenzoate
Purity: 99%
CAS No.: 104901-43-1
Molecular Formula: C9H9BrO2
Formula Weight: 229.07
MDL No.: MFCD00144769
Formula:
C₉H₉BrO₂ MW: 229.07 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00144769 CAS Number: 104901-43-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...