2,3,5,6-Tetrafluoro-4-(trifluoromethyl)aniline 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-Aminotetrafluorobenzotrifluoride
B24326-03
B24326-06
CAAAB24326-03EA
93.54
CAD
CAAAB24326-03
CAAAB24326-06
2,3,5,6-Tetrafluoro-4-(trifluoromethyl)aniline 98%
2,3,5,6-Tetrafluoro-4-(trifluoromethyl)aniline
Purity: 98%
CAS No.: 651-83-2
Molecular Formula: C7H2F7N
Formula Weight: 233.09
Refractive Index: 1.4300
Boiling Point: 185-187°
UN No.: 2810
MDL No.: MFCD00091518
Formula:
C₇H₂F₇N MW: 233.09 g/mol Boiling Pt: 185…187 °C Storage Temperature: Ambient |
MDL Number:
MFCD00091518 CAS Number: 651-83-2 UN: 2810 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...