N-Acetyl-L-methionine 99%
Supplier: Thermo Scientific Chemicals
Synonyms:
N-Ac-Met-OH, N-Acetyl-L-Methionine, Ac-Met-OH NAM, Ac-Met-OH
B24312-06
B24312-14
CAAAB24312-06EA
77.58
CAD
CAAAB24312-06
CAAAB24312-14
CAAAB24312-22
N-Acetyl-L-methionine 99%
N-Acetyl-L-methionine
Purity: 99%
CAS No.: 65-82-7
Molecular Formula: C7H13NO3S
Formula Weight: 191.25
Melting Point: 103-107°
Beilstein Reference: 1725552
Merck Reference: 14,96
Optical Rotation: -21° (c=1.3 in water)
MDL No.: MFCD00064441
Formula:
C₇H₁₃NO₃S MW: 191.2 g/mol Melting Pt: 103…107 °C Storage Temperature: Ambient |
MDL Number:
MFCD00064441 CAS Number: 65-82-7 EINECS: 200-617-7 Merck Index: 14,96 |
Specification Test Results
Optical rotation | -21° (c=1.3 in water) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...