Ethyl-3-(dimethylamino)acrylate 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B24291-09
B24291-18
B24291-30
CAAAB24291-09EA
77.58
CAD
CAAAB24291-09
CAAAB24291-18
CAAAB24291-30
Ethyl-3-(dimethylamino)acrylate 99%
Ethyl-3-(dimethylamino)acrylate
Purity: 99%
CAS No.: 924-99-2
Molecular Formula: C7H13NO2
Formula Weight: 143.19
Refractive Index: 1.5120
Melting Point: 17-18°
Boiling Point: 118-121°/10mm
MDL No.: MFCD00144269
Formula:
C₇H₁₃NO₂ Boiling Pt: 118…121 °C/10 mm Melting Pt: 17…18 °C Storage Temperature: Ambient |
MDL Number:
MFCD00144269 CAS Number: 924-99-2 EINECS: 402-650-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...