Zincon sodium salt
Supplier: Thermo Scientific Chemicals
|
Warning
|
B24256-03
B24256-06
B24256-14
CAAAB24256-03EA
55.77
CAD
CAAAB24256-03
CAAAB24256-06
CAAAB24256-14
Zincon sodium salt
Zincon sodium salt
CAS No.: 62625-22-3
Molecular Formula: C20H15N4NaO6S
Formula Weight: 462.42
Melting Point: ca 203° dec.
Beilstein Reference: 3872699
MDL No.: MFCD00064385
Formula:
C₂₀H₁₅N₄NaO₆S Melting Pt: ca 203 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00064385 CAS Number: 62625-22-3 EINECS: 263-651-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...