2,2-Difluoro-1,3-benzodioxole-5-carboxaldehyde 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B24232-03
B24232-06
CAAAB24232-03EA
114.55
CAD
CAAAB24232-03
CAAAB24232-06
2,2-Difluoro-1,3-benzodioxole-5-carboxaldehyde 97%
2,2-Difluoro-1,3-benzodioxole-5-carboxaldehyde
Purity: 97%
CAS No.: 656-42-8
Molecular Formula: C8H4F2O3
Formula Weight: 186.11
Refractive Index: 1.4990
Boiling Point: 103-105°/20mm
MDL No.: MFCD00792420
Formula:
C₈H₄F₂O₃ Boiling Pt: 103…105 °C/20 mm Density: 1.399 Storage Temperature: Ambient |
MDL Number:
MFCD00792420 CAS Number: 656-42-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...