2-Bromohexanoic acid 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
B24228-14
B24228-22
CAAAB24228-14EA
53.25
CAD
CAAAB24228-14
CAAAB24228-22
2-Bromohexanoic acid 97%
2-Bromohexanoic acid
Purity: 97%
CAS No.: 616-05-7
Molecular Formula: CH3(CH2)3CHBrCO2H
Formula Weight: 195.06
Flash Point:
Formula:
C₆H₁₁BrO₂ Boiling Pt: 136…138 °C/18 mm Melting Pt: 3…5 °C Density: 1.37 Flash Pt: >110 °C(230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004218 CAS Number: 616-05-7 EINECS: 210-461-1 UN: 2922 ADR: "8",III Merck Index: 14,1415 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...