Methyl vanillate 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Methyl 4-hydroxy-3-methoxybenzoate

B24209-06 B24209-14 B24209-22
CAAAB24209-06EA 68.57 CAD
CAAAB24209-06 CAAAB24209-14 CAAAB24209-22
Methyl vanillate 99%
Methyl vanillate

Purity: 99% CAS No.: 3943-74-6 Molecular Formula: C9H10O4 Formula Weight: 182.18 Melting Point: 62-66° Boiling Point: 285-287° MDL No.: MFCD00008438

Formula: C₉H₁₀O₄
Boiling Pt: 285…287 °C
Melting Pt: 62…66 °C
Storage Temperature: Ambient
MDL Number: MFCD00008438
CAS Number: 3943-74-6
EINECS: 223-525-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR