(Chloromethylene)dimethylammonium chloride 96%

Supplier: Thermo Scientific Chemicals
Danger

B24172-06 B24172-14
CAAAAB24172-06EA 76.16 CAD
CAAAAB24172-06 CAAAAB24172-14
(Chloromethylene)dimethylammonium chloride 96%
(Chloromethylene)dimethylammonium chloride

Fieser: 4,186 5,251 6,220 7,422 8,186 9,514 10,457 12,564 13,341


Purity: 96% CAS No.: 3724-43-4 Molecular Formula: C3H7Cl2N Formula Weight: 128.00 Storage Temperature: 2°C to 8°C Odor: Acrid Beilstein Reference: 3566322 UN No.: 3261 MDL No.: MFCD00011868

Formula: C₃H₇Cl₂N
Melting Pt: 130 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00011868
CAS Number: 3724-43-4
EINECS: 425-970-6
UN: 3261
ADR: "8",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR