2-(1-Naphthylmethyl)-2-imidazoline hydrochloride 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Naphazoline Hydrochloride, Albalon
B24161-14
B24161-22
B24161-36
CAAAB24161-14EA
60.04
CAD
CAAAB24161-14
CAAAB24161-22
CAAAB24161-36
2-(1-Naphthylmethyl)-2-imidazoline hydrochloride 99%
2-(1-Naphthylmethyl)-2-imidazoline hydrochloride
Purity: 99%
CAS No.: 550-99-2
Molecular Formula: C14H14N2.HCl
Formula Weight: 246.74
Melting Point: 257-261°
Beilstein Reference: 3716843
Merck Reference: 14,6368
UN No.: 2811
MDL No.: MFCD00012554
Formula:
C₁₄H₁₅ClN₂ Melting Pt: 257…261 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012554 CAS Number: 550-99-2 EINECS: 208-989-2 UN: 2811 ADR: 6.1,III Merck Index: 14,6368 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...