Maltitol 97%
Supplier: Thermo Scientific Chemicals
B24158-0B
B24158-18
B24158-30
CAAAB24158-0BEA
518.24
CAD
CAAAB24158-0B
CAAAB24158-18
CAAAB24158-30
Maltitol 97%
Maltitol
Purity: 97%
CAS No.: 585-88-6
Molecular Formula: C12H24O11
Formula Weight: 344.32
Physical Form: Cryst powder
Appearance: White
Melting Point: 149-152°
Beilstein Reference: 89983
Optical Rotation: +107° (c=10 in water)
MDL No.: MFCD00006600
Formula:
C₁₂H₂₄O₁₁ Melting Pt: 149…152 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006600 CAS Number: 585-88-6 EINECS: 209-567-0 |
Specification Test Results
Optical rotation | +107° (c=10 in water) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...