3-Decanol 97%
Supplier: Thermo Scientific Chemicals
B24141-06
B24141-14
CAAAB24141-06EA
96.54
CAD
CAAAB24141-06
CAAAB24141-14
CAAAB24141-22
3-Decanol 97%
3-Decanol
Purity: 97%
CAS No.: 1565-81-7
Molecular Formula: C10H22O
Formula Weight: 158.29
Flash Point: 99°(210°F)
Refractive Index: 1.4350
Boiling Point: 213°
Beilstein Reference: 1719658
MDL No.: MFCD00021956
Formula:
C₁₀H₂₂O Boiling Pt: 213 °C Density: 0.830 Flash Pt: 99 °C(210 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00021956 CAS Number: 1565-81-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...