Methyl-α-L-rhamnopyranoside 98%

Supplier: Thermo Scientific Chemicals

B24129-06 B24129-14
CAAAB24129-06EA 129.88 CAD
CAAAB24129-06 CAAAB24129-14
Methyl-α-L-rhamnopyranoside 98%
Methyl-alpha-L-rhamnopyranoside

Purity: 98% CAS No.: 14917-55-6 Molecular Formula: C7H14O5 Formula Weight: 178.19 Melting Point: 106-108° MDL No.: MFCD00067654

Formula: C₇H₁₄O₅
Melting Pt: 106…108 °C
Storage Temperature: Ambient
MDL Number: MFCD00067654
CAS Number: 14917-55-6
EINECS: 238-987-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR