Cobalt(II) citrate dihydrate 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
B24088-14
B24088-22
CAAAAB24088-14EA
88.48
CAD
CAAAAB24088-14
CAAAAB24088-22
Cobalt(II) citrate dihydrate 98%
Cobalt(II) citrate dihydrate
Purity: 98%
CAS No.: 6424-15-3
Molecular Formula: C12H10Co3O14.2H2O
Formula Weight: 591.02 (554.99anhy)
Melting Point: ca 150° dec.
UN No.: 3077
MDL No.: MFCD02684007
Formula:
C₁₂H₁₀Co₃O₁₄·2H₂O Melting Pt: ca 150 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD02684007 CAS Number: 6424-15-3 EINECS: 212-751-3 UN: 3077 ADR: "9",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...