(3-Chlorophenyl)methanesulfonyl chloride 96%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
3-Chlorobenzylsulphonyl chloride, 3-Chlorobenzylsulfonyl chloride, 3-Chloro-α-toluenesulphonyl chloride
B24073-03
B24073-06
B24073-14
CAAAB24073-03EA
76.63
CAD
CAAAB24073-03
CAAAB24073-06
CAAAB24073-14
(3-Chlorophenyl)methanesulfonyl chloride 96%
(3-Chlorophenyl)methanesulphonyl chloride
Purity: 96%
CAS No.: 24974-73-0
Molecular Formula: C7H6Cl2O2S
Formula Weight: 225.09
Melting Point: 69-75°
UN No.: 3261
MDL No.: MFCD02683111
Formula:
C₇H₆Cl₂O₂S Melting Pt: 69…75 °C Storage Temperature: Ambient |
MDL Number:
MFCD02683111 CAS Number: 24974-73-0 UN: 3261 ADR: "8",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...