Ferrozine mono-sodium salt hydrate 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B24066-03
B24066-06
B24066-14
CAAAB24066-03EA
75.84
CAD
CAAAB24066-03
CAAAB24066-06
CAAAB24066-14
Ferrozine mono-sodium salt hydrate 97%
Ferrozine mono-sodium salt hydrate
Purity: 97%
CAS No.: 69898-45-9
Molecular Formula: C20H13N4NaO6S2.xH2O
Formula Weight: 492.47(anhy)
Beilstein Reference: 5222087
MDL No.: MFCD00150794
Formula:
C₂₀H₁₃N₄NaO₆S₂·H₂O Storage Temperature: Ambient |
MDL Number:
MFCD00150794 CAS Number: 63451-29-6 EINECS: 274-196-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...