2-(4-Aminophenyl)-1,1,1,3,3,3-hexafluoro-2-propanol 96%

Supplier: Thermo Scientific Chemicals
Warning

B24048-03 B24048-06 B24048-14
CAAAB24048-03EA 48.51 CAD
CAAAB24048-03 CAAAB24048-06 CAAAB24048-14
2-(4-Aminophenyl)-1,1,1,3,3,3-hexafluoro-2-propanol 96%
2-(4-Aminophenyl)-1,1,1,3,3,3-hexafluoro-2-propanol

Purity: 96% CAS No.: 722-92-9 Molecular Formula: C9H7F6NO Formula Weight: 259.15 Melting Point: 147-152° Beilstein Reference: 1473737 MDL No.: MFCD00039258

Formula: C₉H₇F₆NO
Melting Pt: 147…152 °C
Storage Temperature: Ambient
MDL Number: MFCD00039258
CAS Number: 722-92-9
EINECS: 670-530-0
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR