1-(2-Thiazolylazo)-2-naphthol 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B24011-03
B24011-06
B24011-14
CAAAB24011-03EA
56.41
CAD
CAAAB24011-03
CAAAB24011-06
CAAAB24011-14
1-(2-Thiazolylazo)-2-naphthol 98%
1-(2-Thiazolylazo)-2-naphthol
Purity: 98%
CAS No.: 1147-56-4
Molecular Formula: C13H9N3OS
Formula Weight: 255.30
Melting Point: 138-140°
Beilstein Reference: 223783
Formula:
C₁₃H₉N₃OS Melting Pt: 138…140 °C Storage Temperature: Ambient |
MDL Number:
MFCD00021611 CAS Number: 1147-56-4 EINECS: 214-555-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...