Dimethyl-1,4-cyclohexanedione-2,5-dicarboxylate 99+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Dimethyl 2,5-dioxocyclohexane-1,4-dicarboxylate

B24008-22 B24008-36
CAAAB24008-14EA 61.79 CAD
CAAAB24008-14 CAAAB24008-22 CAAAB24008-36
Dimethyl-1,4-cyclohexanedione-2,5-dicarboxylate 99+%
Dimethyl 1,4-cyclohexanedione-2,5-dicarboxylate

Purity: 99+% CAS No.: 6289-46-9 Molecular Formula: C10H12O6 Formula Weight: 228.2 Melting Point: 152-156° MDL No.: MFCD00001607

Formula: C₁₀H₁₂O₆
MW: 228.2 g/mol
Melting Pt: 152…156 °C
Storage Temperature: Ambient
MDL Number: MFCD00001607
CAS Number: 6289-46-9
EINECS: 228-528-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR