3,6-Di-tert-butylfluorene 98%
Supplier: Thermo Scientific Chemicals
B23996-03
B23996-06
CAAAB23996-03EA
188.02
CAD
CAAAB23996-03
CAAAB23996-06
3,6-Di-tert-butylfluorene 98%
3,6-Di-tert-butylfluorene
Purity: 98%
CAS No.: 58775-07-8
Molecular Formula: C21H26
Formula Weight: 278.44
Melting Point: 79-82°
MDL No.: MFCD02683109
Formula:
C₂₁H₂₆ MW: 278.44 g/mol Melting Pt: 79…82 °C Storage Temperature: Ambient |
MDL Number:
MFCD02683109 CAS Number: 58775-07-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...