4-(1-Pyrrolidinyl)pyridine 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 4-Pyrrolidinopyridine

B23989-06 B23989-14 B23989-22
CAAAB23989-06EA 118.34 CAD
CAAAB23989-06 CAAAB23989-14 CAAAB23989-22
4-(1-Pyrrolidinyl)pyridine 98%
4-(1-Pyrrolidinyl)pyridine

Fieser: 4,416 9,400 10,336 13,262


Purity: 98% CAS No.: 2456-81-7 Molecular Formula: C9H12N2 Formula Weight: 148.21 Flash Point: 142°(287°F) Melting Point: 54-58° Boiling Point: 171-173°/21mm Beilstein Reference: 472443 UN No.: 2923 MDL No.: MFCD00006415

Formula: C₉H₁₂N₂
MW: 148.21 g/mol
Boiling Pt: 171…173 °C/21 mm
Melting Pt: 54…58 °C
Flash Pt: 142 °C(287 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006415
CAS Number: 2456-81-7
EINECS: 219-534-2
UN: 2923
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR