1-Amino-4-naphthol hydrochloride tech. 90%, Technical Grade

Supplier: Thermo Scientific Chemicals
Warning

B23978-03 B23978-06 B23978-14
CAAAB23978-03EA 41.55 CAD
CAAAB23978-03 CAAAB23978-06 CAAAB23978-14
1-Amino-4-naphthol hydrochloride tech. 90%, Technical Grade
1-Amino-4-naphthol hydrochloride

Purity: 90% CAS No.: 5959-56-8 Molecular Formula: C10H9NO.HCl Formula Weight: 195.65 Melting Point: ca 272° dec. MDL No.: MFCD00012562

Formula: C₁₀H₁₀ClNO
Melting Pt: ca 272 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012562
CAS Number: 5959-56-8
EINECS: 227-727-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR