2,2'-Furil dioxime mixture of isomers 97%

Supplier: Thermo Scientific Chemicals

B23972-03 B23972-06
CAAAB23972-03EA 172.22 CAD
CAAAB23972-03 CAAAB23972-06
2,2'-Furil dioxime mixture of isomers 97%
2,2'-Furil dioxime

Purity: 97% CAS No.: 522-27-0 Molecular Formula: C10H8N2O4 Formula Weight: 220.19 Melting Point: ca 165° Beilstein Reference: 85894 MDL No.: MFCD00075315

Formula: C₁₀H₈N₂O₄
MW: 220.19
Melting Pt: ∼ 165 °C
Storage Temperature: Ambient
MDL Number: MFCD00075315
CAS Number: 522-27-0
EINECS: 208-326-7
Merck Index: 14,4306

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR