2-(Bromoacetyl)naphthalene 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
B23960-09
B23960-18
CAAAB23960-09EA
112.34
CAD
CAAAB23960-09
CAAAB23960-18
CAAAB23960-30
2-(Bromoacetyl)naphthalene 98%
2-(Bromoacetyl)naphthalene
Purity: 98%
CAS No.: 613-54-7
Molecular Formula: C12H9BrO
Formula Weight: 249.11
Melting Point: 80-85°
Beilstein Reference: 639153
UN No.: 3261
MDL No.: MFCD00004109
Formula:
C₁₂H₉BrO Melting Pt: 80…85 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004109 CAS Number: 613-54-7 EINECS: 210-348-7 UN: 3261 ADR: "8",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...