Iron(II) fumarate 94%
Supplier: Thermo Scientific Chemicals
B23956-0B
B23956-30
CAAAB23956-0BEA
218.04
CAD
CAAAB23956-0B
CAAAB23956-30
Iron(II) fumarate 94%
Iron(II) fumarate
Purity: 94%
CAS No.: 141-01-5
Molecular Formula: C4H2FeO4
Formula Weight: 169.90
Melting Point:
Formula:
C₄H₂FeO₄ Melting Pt: >300 °C Density: 2.44 Storage Temperature: Ambient |
MDL Number:
MFCD00058315 CAS Number: 141-01-5 EINECS: 205-447-7 Merck Index: 14,4046 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...