2,2-Difluoro-1,3-benzodioxole-4-carboxylic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B23955-03
B23955-06
CAAAB23955-03EA
73.79
CAD
CAAAB23955-03
CAAAB23955-06
2,2-Difluoro-1,3-benzodioxole-4-carboxylic acid 97%
2,2-Difluoro-1,3-benzodioxole-4-carboxylic acid
Purity: 97%
CAS No.: 126120-85-2
Molecular Formula: C8H4F2O4
Formula Weight: 202.12
Melting Point: 202-205°
Beilstein Reference: 7292629
Formula:
C₈H₄F₂O₄ Melting Pt: 202…205 °C Storage Temperature: Ambient |
MDL Number:
MFCD01631473 CAS Number: 126120-85-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...