meso-Hydrobenzoin 95%
Supplier: Thermo Scientific Chemicals
B23946-06
B23946-14
CAAAB23946-06EA
112.18
CAD
CAAAB23946-06
CAAAB23946-14
meso-Hydrobenzoin 95%
meso-Hydrobenzoin
Purity: 99%
CAS No.: 579-43-1
Molecular Formula: C14H14O2
Formula Weight: 214.27
Melting Point: 134-136°
Beilstein Reference: 2050814
MDL No.: MFCD00064253
Formula:
C₁₄H₁₄O₂ Melting Pt: 134…136 °C Storage Temperature: Ambient |
MDL Number:
MFCD00064253 CAS Number: 579-43-1 EINECS: 207-758-3 Merck Index: 14,4777 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...