4-(Diethylamino)azobenzene 98%

Supplier: Thermo Scientific Chemicals
Warning

B23915-06 B23915-14 B23915-22
CAAAB23915-06EA 57.51 CAD
CAAAB23915-06 CAAAB23915-14 CAAAB23915-22
4-(Diethylamino)azobenzene 98%
4-(Diethylamino)azobenzene

Purity: 98% CAS No.: 2481-94-9 Molecular Formula: C16H19N3 Formula Weight: 253.35 Melting Point: 93-96° MDL No.: MFCD00009043

Formula: C₁₆H₁₉N₃
Melting Pt: 93…96 °C
Storage Temperature: Ambient
MDL Number: MFCD00009043
CAS Number: 2481-94-9
EINECS: 219-616-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR