2-Amino-4-(1-naphthyl)thiazole 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-(Naphthalen-1-yl)thiazol-2-amine
B23882-03
B23882-06
CAAAB23882-03EA
87.22
CAD
CAAAB23882-03
CAAAB23882-06
2-Amino-4-(1-naphthyl)thiazole 97%
2-Amino-4-(1-naphthyl)thiazole
Purity: 97%
CAS No.: 56503-96-9
Molecular Formula: C13H10N2S
Formula Weight: 226.30
Melting Point: 155-158°
Beilstein Reference: 164858
Formula:
C₁₃H₁₀N₂S MW: 226.30 g/mol Melting Pt: 155…158 °C Storage Temperature: Ambient |
MDL Number:
MFCD00236016 CAS Number: 56503-96-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...