tert-Butylhydrazine hydrochloride 98%

Supplier: Thermo Scientific Chemicals
Warning

B23876-14 B23876-22 B23876-36
CAAAB23876-14EA 45.98 CAD
CAAAB23876-14 CAAAB23876-22 CAAAB23876-36
tert-Butylhydrazine hydrochloride 98%
tert-Butylhydrazine hydrochloride

Purity: 98% CAS No.: 7400-27-3 Molecular Formula: (CH3)3CNHNH2.HCl Formula Weight: 124.61 Melting Point: ca 192° dec. Beilstein Reference: 3651224

Formula: C₄H₁₃ClN₂
Melting Pt: ca 192 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012947
CAS Number: 7400-27-3
EINECS: 231-001-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR