(3-Acetamidophenyl)boronic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Acetamidobenzeneboronic acid, 3-Acetamidophenylboronic acid

B23833-03 B23833-06
CAAAB23833-03EA 115.97 CAD
CAAAB23833-03 CAAAB23833-06 CAAAB23833-14
(3-Acetamidophenyl)boronic acid 98%
(3-Acetamidophenyl)boronic acid

Purity: 98% CAS No.: 78887-39-5 Molecular Formula: C8H10BNO3 Formula Weight: 178.98 Melting Point: ca 263° dec. Beilstein Reference: 3278316

Formula: C₈H₁₀BNO₃
MW: 178.98 g/mol
Melting Pt: ∼ 263 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00236013
CAS Number: 78887-39-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR