N-Acetyl-L-hydroxyproline 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Ac-Hyp-OH
B23767-04
B23767-09
CAAAB23767-04EA
75.05
CAD
CAAAB23767-04
CAAAB23767-09
CAAAB23767-18
N-Acetyl-L-hydroxyproline 99%
N-Acetyl-L-hydroxyproline
Purity: 99%
CAS No.: 33996-33-7
Molecular Formula: C7H11NO4
Formula Weight: 173.17
Melting Point: 131-133°
Beilstein Reference: 84043
Optical Rotation: -117° (c=2 in water)
MDL No.: MFCD00037339
Formula:
C₇H₁₁NO₄ MW: 173.168 g/mol Melting Pt: 131…133 °C Storage Temperature: Ambient |
MDL Number:
MFCD00037339 CAS Number: 33996-33-7 EINECS: 251-780-6 Merck Index: 14,6903 |
Specification Test Results
Optical rotation | -117° (c=2 in water) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...