6-Hydroxyflavanone 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B23755-03
B23755-06
CAAAB23755-03EA
74.42
CAD
CAAAB23755-03
CAAAB23755-06
CAAAB23755-14
6-Hydroxyflavanone 98+%
6-Hydroxyflavanone
Purity: 98+%
CAS No.: 4250-77-5
Molecular Formula: C15H12O3
Formula Weight: 240.25
Melting Point: ca 220° dec.
Beilstein Reference: 87354
Formula:
C₁₅H₁₂O₃ Melting Pt: ca 220 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00017485 CAS Number: 4250-77-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...