Tetrakis(dimethylsiloxy)silane 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B23754-06
B23754-14
B23754-22
CAAAB23754-06EA
57.99
CAD
CAAAB23754-06
CAAAB23754-14
CAAAB23754-22
Tetrakis(dimethylsiloxy)silane 97%
Tetrakis(dimethylsiloxy)silane
Purity: 97%
CAS No.: 17082-47-2
Molecular Formula: C8H28O4Si5
Formula Weight: 328.73
Flash Point: 67°(153°F)
Refractive Index: 1.3880
Melting Point: -60°
Boiling Point: 188-190°
Beilstein Reference: 1793898
MDL No.: MFCD00053848
Formula:
C₈H₂₈O₄Si₅ Boiling Pt: 188…190 °C Melting Pt: –60 °C Density: 0.884 Flash Pt: 67 °C(153 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00053848 CAS Number: 17082-47-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...