(±)-Diethyl acetylsuccinate 97+%

Supplier: Thermo Scientific Chemicals

Synonyms: Diethyl 2-acetylsuccinate, Diethyl acetylsuccinate

B23744-06 B23744-14 B23744-22
CAAAB23744-06EA 47.08 CAD
CAAAB23744-06 CAAAB23744-14 CAAAB23744-22
(±)-Diethyl acetylsuccinate 97+%
(±)-Diethyl acetylsuccinate

Purity: 97+% CAS No.: 1115-30-6 Molecular Formula: C10H16O5 Formula Weight: 216.23 Flash Point: 110°(230°F) Refractive Index: 1.4346 Melting Point: -8° Boiling Point: 180-183°/50mm MDL No.: MFCD00009157

Formula: C₁₀H₁₆O₅
MW: 216.23 g/mol
Boiling Pt: 180…183 °C/50 mm
Melting Pt: –8 °C
Density: 1.081
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009157
CAS Number: 1115-30-6
EINECS: 214-223-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR