3-[(Trifluoromethyl)thio]aniline 96%

Supplier: Thermo Scientific Chemicals
Warning

B23734-03 B23734-06
CAAAB23734-03EA 133.19 CAD
CAAAB23734-03 CAAAB23734-06
3-[(Trifluoromethyl)thio]aniline 96%
3-[(Trifluoromethyl)thio]aniline

Purity: 96% CAS No.: 369-68-6 Molecular Formula: C7H6F3NS Formula Weight: 193.19 Flash Point: 103°(218°F) Refractive Index: 1.5220 Odor: Unpleasant Boiling Point: 221-224° UN No.: 2810 MDL No.: MFCD00190130

Formula: C₇H₆F₃NS
MW: 193.19
Boiling Pt: 221…224 °C
Density: 1.348
Flash Pt: 103 °C(218 °F)
Storage Temperature: Ambient
MDL Number: MFCD00190130
CAS Number: 369-68-6
EINECS: 206-722-4
UN: 2810
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR