Ethyl-2,2-diethylacetoacetate 97%
Supplier: Thermo Scientific Chemicals
B23729-06
B23729-14
CAAAB23729-06EA
69.99
CAD
CAAAB23729-06
CAAAB23729-14
Ethyl-2,2-diethylacetoacetate 97%
Ethyl-2,2-diethylacetoacetate
Purity: 97%
CAS No.: 1619-57-4
Molecular Formula: C10H18O3
Formula Weight: 186.25
Refractive Index: 1.4330
Boiling Point: 84-86°/13mm
Beilstein Reference: 1767891
MDL No.: MFCD00048552
Formula:
C₁₀H₁₈O₃ MW: 186.25 g/mol Boiling Pt: 84…86 °C/13 mm Storage Temperature: Ambient |
MDL Number:
MFCD00048552 CAS Number: 1619-57-4 EINECS: 216-581-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...