4-Biphenylboronic acid 97+%

Supplier: Thermo Scientific Chemicals
Warning

B23703-03 B23703-06 B23703-14
CAAAB23703-03EA 55.93 CAD
CAAAB23703-03 CAAAB23703-06 CAAAB23703-14
4-Biphenylboronic acid 97+%
4-Biphenylboronic acid

Purity: 98+% CAS No.: 5122-94-1 Molecular Formula: C12H11BO2 Formula Weight: 198.03 Odor: Odorless Melting Point: 238-247° Beilstein Reference: 2937410 MDL No.: MFCD00093311

Formula: C₁₂H₁₁BO₂
Melting Pt: 238…247 °C
Storage Temperature: Ambient
MDL Number: MFCD00093311
CAS Number: 5122-94-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR