2-Methoxy-5-sulfamoylbenzoic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

B23698-09 B23698-18
CAAAB23698-09EA 35.87 CAD
CAAAB23698-09 CAAAB23698-18 CAAAB23698-30
2-Methoxy-5-sulfamoylbenzoic acid 97%
2-Methoxy-5-sulphamoylbenzoic acid

Purity: 97% CAS No.: 22117-85-7 Molecular Formula: C8H9NO5S Formula Weight: 232.23 MDL No.: MFCD00129997

Formula: C₈H₉NO₅S
MW: 232.23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00129997
CAS Number: 22117-85-7
EINECS: 244-789-1

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR