Diethoxymethylphenylsilane 97+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B23684-09
B23684-18
CAAAB23684-09EA
38.87
CAD
CAAAB23684-09
CAAAB23684-18
Diethoxymethylphenylsilane 97+%
Diethoxymethylphenylsilane
Purity: 97+%
CAS No.: 775-56-4
Molecular Formula: (CH3CH2O)2Si(CH3)C6H5
Formula Weight: 210.35
Flash Point: 88°(190°F)
Refractive Index: 1.4696
Boiling Point: 117-118°/30mm
Beilstein Reference: 2938396
MDL No.: MFCD00048541
Formula:
C₁₁H₁₈O₂Si Boiling Pt: 117…118 °C/30 mm Density: 0.963 Flash Pt: 88 °C(190 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00048541 CAS Number: 775-56-4 EINECS: 212-275-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...