Nickel(II) lactate tetrahydrate 98%

Supplier: Thermo Scientific Chemicals
Danger

B23643-14 B23643-22 B23643-36
CAAAB23643-14EA 44.4 CAD
CAAAB23643-14 CAAAB23643-22 CAAAB23643-36
Nickel(II) lactate tetrahydrate 98%
Nickel(II) lactate tetrahydrate

Purity: 98% Molecular Formula: C6H10NiO6.4H2O Formula Weight: 308.91 (236.85anhy) UN No.: 3077 MDL No.: MFCD02683106

Formula: C₆H₁₀NiO₆·4H₂O
Storage Temperature: Ambient
MDL Number: MFCD02683106
UN: 3077
ADR: 9,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR