2-Nitrobenzenesulfenyl chloride 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
B23627-09
B23627-18
CAAAB23627-09EA
51.82
CAD
CAAAB23627-09
CAAAB23627-18
2-Nitrobenzenesulfenyl chloride 97%
2-Nitrobenzenesulphenyl chloride
Fieser: 1,745 4,359 6,419 8,360 10,277 11,360 15,217
Purity: 97%
CAS No.: 7669-54-7
Molecular Formula: C6H4ClNO2S
Formula Weight: 189.62
Storage Temperature: 2°C to 8°C
Melting Point: 72-75°
Beilstein Reference: 473741
UN No.: 3261
MDL No.: MFCD00007128
Formula:
C₆H₄ClNO₂S Melting Pt: 72…75 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007128 CAS Number: 7669-54-7 EINECS: 231-644-2 UN: 1759 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...