6-Acetamidohexanoic acid hydrate 99%

Supplier: Thermo Scientific Chemicals

B23623-06 B23623-14
CAAAB23623-06EA 52.3 CAD
CAAAB23623-06 CAAAB23623-14
6-Acetamidohexanoic acid hydrate 99%
6-Acetamidohexanoic acid hydrate

Purity: 99% CAS No.: 57-08-9 Molecular Formula: C8H15NO3.xH2O Formula Weight: 173.21(anhy) Melting Point: 102-106° MDL No.: MFCD00004424

Formula: C₈H₁₅NO₃·xH₂O
Melting Pt: 102…106 °C
Storage Temperature: Ambient
MDL Number: MFCD00004424
CAS Number: 57-08-9
EINECS: 200-310-8
Merck Index: 14,45

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR