4-Amino-2,5-difluorobenzonitrile 96%

Supplier: Thermo Scientific Chemicals
Danger

B23611-03 B23611-06
CAAAB23611-03EA 50.4 CAD
CAAAB23611-03 CAAAB23611-06
4-Amino-2,5-difluorobenzonitrile 96%
4-Amino-2,5-difluorobenzonitrile

Purity: 96% CAS No.: 112279-61-5 Molecular Formula: C7H4F2N2 Formula Weight: 154.12 Melting Point: 91-94° UN No.: 3439 MDL No.: MFCD00190102

Formula: C₇H₄F₂N₂
MW: 154.12 g/mol
Melting Pt: 91…94 °C
Storage Temperature: Ambient
MDL Number: MFCD00190102
CAS Number: 112279-61-5
UN: 3439
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR