2,2-Difluoro-1,3-benzodioxole-5-carboxylic acid 97+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2,2-Difluorobenzo[d][1,3]dioxole-5-carboxylic acid

B23610-03 B23610-06
CAAAAB23610-03EA 100.65 CAD
CAAAAB23610-03 CAAAAB23610-06
2,2-Difluoro-1,3-benzodioxole-5-carboxylic acid 97+%
2,2-Difluoro-1,3-benzodioxole-5-carboxylic acid

Purity: 97+% CAS No.: 656-46-2 Molecular Formula: C8H4F2O4 Formula Weight: 202.12 Melting Point: 154-157° MDL No.: MFCD00792417

Formula: C₈H₄F₂O₄
MW: 202.12 g/mol
Melting Pt: 154…157 °C
Storage Temperature: Ambient
MDL Number: MFCD00792417
CAS Number: 656-46-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR