4-(Dimethylamino)-2-methoxybenzaldehyde 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B23598-03
B23598-06
CAAAB23598-03EA
192.76
CAD
CAAAB23598-03
CAAAB23598-06
4-(Dimethylamino)-2-methoxybenzaldehyde 98%
4-(Dimethylamino)-2-methoxybenzaldehyde
Purity: 98%
CAS No.: 84562-48-1
Molecular Formula: C10H13NO2
Formula Weight: 179.22
Melting Point: 58-60°
Beilstein Reference: 2719181
Formula:
C₁₀H₁₃NO₂ Melting Pt: 58…60 °C Storage Temperature: Ambient |
MDL Number:
MFCD00151814 CAS Number: 84562-48-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...