HBTU(O-(1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium hexafluorophosphate) 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
HBTU, 1-(Bis(dimethylamino)methylene)-1H-benzo[d][1,2,3]triazole-1-ium 3-oxide hexafluorophosphate(V)
B23597-06
B23597-14
B23597-22
CAAAAB23597-06EA
107.76
CAD
CAAAAB23597-06
CAAAAB23597-14
CAAAAB23597-22
HBTU(O-(1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium hexafluorophosphate) 98%
HBTU (O-(1H-Benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium hexafluorophosphate)
Purity: 98%
CAS No.: 94790-37-1
Molecular Formula: C11H16F6N5OP
Formula Weight: 379.25
Storage Temperature: 2°C to 8°C
Melting Point: ca 200° dec.
Beilstein Reference: 7328329
MDL No.: MFCD00075445
Formula:
C₁₁H₁₆F₆N₅OP Melting Pt: ca 200 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00075445 CAS Number: 94790-37-1 EINECS: 423-020-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...