Ethyl-2-hydroxyisobutyrate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B23592-22
B23592-36
CAAAB23592-22EA
139.36
CAD
CAAAB23592-22
CAAAB23592-36
Ethyl-2-hydroxyisobutyrate 98%
Ethyl 2-hydroxyisobutyrate
Purity: 98%
CAS No.: 80-55-7
Molecular Formula: C6H12O3
Formula Weight: 132.16
Flash Point: 44°(111°F)
Refractive Index: 1.4080
Boiling Point: 149-150°
Beilstein Reference: 878308
UN No.: 3272
MDL No.: MFCD00004458
Formula:
C₆H₁₂O₃ Boiling Pt: 149…150 °C Density: 0.980 Flash Pt: 44 °C(111 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004458 CAS Number: 80-55-7 EINECS: 201-290-3 UN: 3272 ADR: "3",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...