ABD-F [=4-(Aminosulfonyl)-7-fluoro-2,1,3-benzoxadiazole] 98%

Supplier: Thermo Scientific Chemicals

Synonyms: ABD-F, 7-Fluoro-2,1,3-Benzoxadiazole-4-sulfonamide, 4-Fluoro-7-sulfobenzofurazan, ammonium salt, 7-Fluoro-2,1,3-Benzoxadiazole-4-sulphonamide, 4-(Aminosulphonyl)-7-fluoro-2,1,3-benzoxadiazole, 4-(Aminosulfonyl)-7-fluoro-2,1,3-benzoxadiazole, 4-(Aminosulfonyl)-7-fluorobenzofurazane, 7-Fluoro-2,1,3-benzoxadiazole-4-sulfonamide, 7-Fluorobenzofurazan-4-sulfonamide

B23586-MB B23586-MC
CAAAB23586-MBEA 692.04 CAD
CAAAB23586-MB CAAAB23586-MC
ABD-F [=4-(Aminosulfonyl)-7-fluoro-2,1,3-benzoxadiazole] 98%
ABD-F [=4-(Aminosulfonyl)-7-fluoro-2,1,3-benzoxadiazole]

Purity: 98% CAS No.: 91366-65-3 Molecular Formula: C6H4FN3O3S Formula Weight: 217.17 Melting Point: 143-147° MDL No.: MFCD00036687

Formula: C₆H₄FN₃O₃S
MW: 217.17 g/mol
Melting Pt: 143…147 °C
Storage Temperature: Ambient
MDL Number: MFCD00036687
CAS Number: 91366-65-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR